Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB392864 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392864 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392864 Supplier Novus Biologicals Supplier No. NBP26884625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HES-1 Polyclonal specifically detects HES-1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen HES-1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias BHLHB39, bHLHb39hairy homolog (Drosophila), Class B basic helix-loop-helix protein 39, FLJ20408, Hairy and enhancer of split 1, hairy and enhancer of split 1, (Drosophila), Hairy homolog, Hairy-like protein, Hes1, HES-1, hHL, HL, HRYHHL, transcription factor HES-1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: PPLVPIPGGAAPPPGGAPCKLGSQAGEAAKVFGGFQVVPAPDGQFAFLIPNGAFAHSGPVIPVYTSNSG
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3280
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.