Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB005395 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB005395 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB005395 Supplier Novus Biologicals Supplier No. NBP227110JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HES2 Polyclonal specifically detects HES2 in Human, Mouse, Rat, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HES2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BHLHB40, bHLHb40Class B basic helix-loop-helix protein 40, Hairy and enhancer of split 2, hairy and enhancer of split 2 (Drosophila), transcription factor HES-2
Gene Symbols HES2
Host Species Rabbit
Immunogen 8 GDAAELRkSLkPLLEkRR 25
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.