Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP227110SS
Description
HES2 Polyclonal specifically detects HES2 in Human, Mouse, Rat, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HES2 | |
Polyclonal | |
Unconjugated | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
HES2 | |
8 GDAAELRkSLkPLLEkRR 25 | |
0.025 mg | |
DNA replication Transcription Translation and Splicing | |
54626 | |
Human, Mouse, Rat, Primate | |
IgG |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
Immunohistochemistry-Paraffin 5ug/ml | |
BHLHB40, bHLHb40Class B basic helix-loop-helix protein 40, Hairy and enhancer of split 2, hairy and enhancer of split 2 (Drosophila), transcription factor HES-2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Chimpanzee, Human, Mouse, Rat. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction