Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | HES2 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Immunohistochemistry-Paraffin 5ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227110SS
![]() |
Novus Biologicals
NBP227110SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227110
![]() |
Novus Biologicals
NBP227110 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
HES2 Polyclonal specifically detects HES2 in Human, Mouse, Rat, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HES2 | |
Immunohistochemistry-Paraffin 5ug/ml | |
Polyclonal | |
Rabbit | |
DNA replication Transcription Translation and Splicing | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
54626 | |
8 GDAAELRkSLkPLLEkRR 25 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Primate | |
BHLHB40, bHLHb40Class B basic helix-loop-helix protein 40, Hairy and enhancer of split 2, hairy and enhancer of split 2 (Drosophila), transcription factor HES-2 | |
HES2 | |
IgG | |
Affinity Purified | |
Chimpanzee, Human, Mouse, Rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title