Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES6 Antibody (OTI1D6), FITC, Novus Biologicals™
SDP

Catalog No. NB288445 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288445 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288445 Supplier Novus Biologicals Supplier No. NBP272008F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HES6 Monoclonal antibody specifically detects HES6 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen HES6
Applications Western Blot
Classification Monoclonal
Clone OTI1D6
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias BHLHB41, bHLHb41Class B basic helix-loop-helix protein 41, bHLHc23, C-HAIRY1, Hairy and enhancer of split 6, hairy and enhancer of split 6 (Drosophila), HES-6, transcription cofactor HES-6
Host Species Mouse
Immunogen Full length human recombinant protein of human HES6 (NP_061115) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 55502
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.