Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227417 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227417 0.1 mg
NBP227417SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227417 Supplier Novus Biologicals Supplier No. NBP227417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HES7 Polyclonal specifically detects HES7 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen HES7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 2-6ug/ml
Formulation No buffer with No Preservative
Gene Alias bHLH factor Hes7, BHLHB37, bHLHb37transcription factor HES-7, Class B basic helix-loop-helix protein 37, Hairy and enhancer of split 7, hairy and enhancer of split 7 (Drosophila), hHes7, SCDO4
Gene Symbols HES7
Host Species Rabbit
Immunogen RAENRDGPkMLkPLVEkRc (aa 6-23)
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84667
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.