Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HES7 Rabbit anti-Human, Alexa Fluor 750, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB043308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB043308 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB043308 Supplier Novus Biologicals Supplier No. NBP297427AF750

Rabbit Polyclonal Antibody

HES7 Polyclonal specifically detects HES7 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HES7
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias bHLH factor Hes7, BHLHB37, bHLHb37transcription factor HES-7, Class B basic helix-loop-helix protein 37, Hairy and enhancer of split 7, hairy and enhancer of split 7 (Drosophila), hHes7, SCDO4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human HES7 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84667
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.