Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexacosanoic acid, 98%

Catalog No. p-141614
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle

CAS: 506-46-7 | C26H52O2 | 396.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-46-7
Molecular Formula C26H52O2
Molecular Weight (g/mol) 396.7
MDL Number MFCD00002811
InChI Key XMHIUKTWLZUKEX-UHFFFAOYSA-N
Synonym cerotic acid, ceric acid, ceratinic acid, cerinic acid, hexacosanic acid, cerylic acid, cerotinic acid, n-hexacosanoic acid, unii-d42cqn6p36, cerate
PubChem CID 10469
ChEBI CHEBI:31009
IUPAC Name hexacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 87.0°C to 89.0°C
Color White
Density 0.8100g/mL
Boiling Point 250.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4301
Linear Formula CH3(CH2)24COOH
Quantity 100 mg
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene,ether and acetone
Formula Weight 396.7
Percent Purity 98%
Physical Form Shiny Fluffy Crystalline Powder
Chemical Name or Material Hexacosanoic acid, 98%
Show More Show Less
EINECSNumber 208-040-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.