Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecane (Certified ACS), Fisher Chemical

Catalog No. o3035500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.448
MDL Number MFCD00008998
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
Synonym n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
PubChem CID 11006
ChEBI CHEBI:45296
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC

Specifications

Melting Point 18°C
Color Colorless
Density 0.773g/cm³
Boiling Point 287°C
Flash Point 135°C
Packaging Glass Bottle
Linear Formula CH3(CH2)14CH3
Quantity 500 mL
Formula Weight 226.45
Percent Purity ≥98%
Grade Certified
Physical Form Liquid
Chemical Name or Material Hexadecane
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes and skin. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause an allergic skin reaction. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:1
Instability:0
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.