Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexafluorobenzene, 99%

Catalog No. aaa1150030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1150030 250 g
AAA1150009 10 g
AAA1150018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1150030 Supplier Thermo Scientific Chemicals Supplier No. A1150030
Only null left
Add to Cart

CAS: 392-56-3 | C6F6 | 186.056 g/mol

Hexafluorobenzene is used as a solvent in photochemical reactions. It is also used as a reference compound in fluorine-19 NMR, carbon-13 NMR. It is used as a solvent in proton NMR, IR spectrum and UV-spectra. It is used as anticorrosive, antifriction and anti-tumor agents. Further, it is used as a reference molecule to investigate tissue oxygenation studies. It forms series of 1:1 complexes with naphthalene, anthracene, phenanthrene, pyrene and triphenylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexafluorobenzene is used as a solvent in photochemical reactions. It is also used as a reference compound in fluorine-19 NMR, carbon-13 NMR. It is used as a solvent in proton NMR, IR spectrum and UV-spectra. It is used as anticorrosive, antifriction and anti-tumor agents. It forms series of 1:1 complexes with naphthalene, anthracene, phenanthrene, pyrene and triphenylene.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-56-3
Molecular Formula C6F6
Molecular Weight (g/mol) 186.056
MDL Number MFCD00000288
InChI Key ZQBFAOFFOQMSGJ-UHFFFAOYSA-N
Synonym hexafluorobenzene, perfluorobenzene, benzene, hexafluoro, hexafluorbenzol, unii-cmc18t611k, benzene, 1,2,3,4,5,6-hexafluoro, hexa fluorobenzene, hexafluoro benzene, pubchem18879, acmc-1bmus
PubChem CID 9805
ChEBI CHEBI:38589
IUPAC Name 1,2,3,4,5,6-hexafluorobenzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)F

Specifications

Melting Point 4°C to 5°C
Density 1.616
Boiling Point 80°C to 81°C
Flash Point 10°C (50°F)
Odor Aromatic
Refractive Index 1.378
Quantity 250 g
UN Number UN1993
Beilstein 1683438
Merck Index 14,4686
Solubility Information Immiscible with water.
Formula Weight 186.06
Percent Purity 99%
Chemical Name or Material Hexafluorobenzene
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H319;
Highly flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-876-2
RTECSNumber DA3050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.