Learn More
Hexamethylenetetramine Buffer for Hach EZ1001 Aluminium Analyser, Reagents
Specifications
Specifications
| Chemical Name or Material | Buffer |
| CAS | 7732-18-5 |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 4 L |
| Packaging | Poly Bottle |
| Sterility | Non-sterile |
| Solubility Information | Completely miscible |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.