Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanamide, 98%, Thermo Scientific™

Catalog No. p-142096
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 628-02-4
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00008046
InChI Key ALBYIUDWACNRRB-UHFFFAOYSA-N
Synonym capronamide, hexamide, hexylamide, n-caproamide, caproamide, n-hexanamide, hexamide van, hexanoamide, n-capronamide, n-caproic amide
PubChem CID 12332
IUPAC Name hexanamide
SMILES CCCCCC(=O)N

Specifications

Melting Point 100.0°C to 102.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)4CONH2
Quantity 1 g
Formula Weight 115.18
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material Hexanamide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-024-8
RTECSNumber MN7875000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.