Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexanes, 95+%, for spectroscopy, mixture of isomers
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
Specifications
Specifications
| Chemical Name or Material | Hexanes |
| Acidity | 0.00015 meq/g max. |
| CAS | 92112-69-1, 110-54-3 |
| Absorbance | (1 cm cell vs water), 0.005 max. at 240nm, 0.005 max. at 250nm, 0.01 max. at 230nm, 0.025 max. at 220nm, 0.1 max. at 210nm, 0.3 max. at 200nm |
| Melting Point | -95°C |
| Density | 0.6590g/mL |
| Boiling Point | 69°C |
| Flash Point | -22°C |
| Assay Percent Range | (95% n-hexane approximately) (GC) |
| Packaging | Glass bottle |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction