Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanoyl-Lysine adduct Antibody (5E8), HRP, Novus Biologicals™
SDP

Catalog No. NBP259363H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259363H 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259363H Supplier Novus Biologicals Supplier No. NBP259363H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hexanoyl-Lysine adduct Monoclonal specifically detects Hexanoyl-Lysine adduct in Non-species specific samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Hexanoyl-Lysine adduct
Applications Western Blot, ELISA
Classification Monoclonal
Clone 500000000
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS with No Preservative
Host Species Mouse
Immunogen Synthetic Hexanoyl modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Test Specificity Specific for Hexanoyl-Lysine adduct (HEL) modified peptides and proteins. Does not detect free Hexanoyl-Lysine. Does not cross-react with Acrolein, Crotonaldehyde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.