Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexaphenylcyclotrisiloxane, 98+%

Catalog No. p-7026818
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2306106

Supplier: Thermo Scientific Chemicals B2306106

Only null left
Add to Cart

CAS: 512-63-0 | C36H30O3Si3 | 594.888 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 512-63-0
Molecular Formula C36H30O3Si3
Molecular Weight (g/mol) 594.888
MDL Number MFCD00023110
InChI Key VCYDUTCMKSROID-UHFFFAOYSA-N
PubChem CID 68187
IUPAC Name 2,2,4,4,6,6-hexakis-phenyl-1,3,5,2,4,6-trioxatrisilinane
SMILES C1=CC=C(C=C1)[Si]2(O[Si](O[Si](O2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6)C7=CC=CC=C7

Specifications

Boiling Point 300°C (1mmHg)
Melting Point ∼175°C to 185°C
Quantity 5 g
Beilstein 2957773
Synonym hexaphenylcyclotrisiloxane, cyclotrisiloxane, hexaphenyl, diphenylsiloxane cyclic trimer, cyclotrisiloxane, 2,2,4,4,6,6-hexaphenyl, acmc-209kso, hexaphenyl-1,3,5,2,4,6-trioxatrisilinane, cyclotrisiloxane,2,2,4,4,6,6-hexaphenyl, 2,2,4,4,6,6-hexaphenyl-1,3,5,2,4,6 trioxatrisilinan, 2,2,4,4,6,6-hexaphenyl-1,3,5,2,4,6-trioxatrisilinane, 2,2,4,4,6,6-hexaphenyl-1,3,5,2,4,6-trioxatrisilinane #
Formula Weight 594.9
Percent Purity ≥98%
Sensitivity Moisture sensitive
Chemical Name or Material Hexaphenylcyclotrisiloxane
EINECSNumber 208-145-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.