Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEXO Antibody (OTI1B3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP272312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272312 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272312 Supplier Novus Biologicals Supplier No. NBP272312
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HEXO Monoclonal specifically detects HEXO in Human, Rat, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen HEXO
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1B3
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Immunohistochemistry 1:150
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias 3′ exoribonuclease, 3′-5′ exonuclease ERI1, 3′EXO, 3′HEXO, EC 3.1, enhanced RNAi three prime mRNA exonuclease homolog 1, Eri-1 homolog, exoribonuclease 1, HEXO, histone mRNA 3′ end-specific exonuclease, Histone mRNA 3′-end-specific exoribonuclease, Histone mRNA 3′-exonuclease 1, MGC35395, Protein 3′hExo, THEX1, three prime histone mRNA exonuclease 1,3′-5′ exoribonuclease 1, three prime mRNA exonuclease 1
Gene Symbols ERI1
Host Species Mouse
Immunogen Full length human recombinant protein of human ERI1(NP_699163) produced in HEK293T cell.
Molecular Weight of Antigen 39.9 kDa
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 90459
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Rat, Primate, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.