Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexokinase 2 Antibody (1A7), FITC, Novus Biologicals™
SDP

Catalog No. NBP104296F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104296F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104296F Supplier Novus Biologicals Supplier No. NBP104296F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hexokinase 2 Monoclonal specifically detects Hexokinase 2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Hexokinase 2
Applications ELISA
Classification Monoclonal
Clone 1A7
Conjugate FITC
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase
Gene Symbols HK2
Host Species Mouse
Immunogen Recombinant human Hexokinase 2 (1-917aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3099
Test Specificity Anti-human Hexokinase 2 mAb, clone 1A7, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Hexokinase 2 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.