Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexosaminidase A/HEXA Antibody (20F1), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N242629APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N242629APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N242629APC7 Supplier Novus Biologicals Supplier No. NBP242629APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hexosaminidase A/HEXA Monoclonal specifically detects Hexosaminidase A/HEXA in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Hexosaminidase A/HEXA
Applications Flow Cytometry
Classification Monoclonal
Clone 20F1
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias beta-hexosaminidase subunit alpha, Beta-N-acetylhexosaminidase subunit alpha, EC 3.2.1, EC 3.2.1.52, hexosaminidase A (alpha polypeptide), Hexosaminidase subunit A, MGC99608, N-acetyl-beta-glucosaminidase subunit alpha, TSD
Gene Symbols HEXA
Host Species Mouse
Immunogen Recombinant human HEXA (89-529aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3073.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.