Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexosaminidase A/HEXA Antibody (20F1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP242629 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242629 Supplier Novus Biologicals Supplier No. NBP242629
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, ELISA, Flow Cytometry

Hexosaminidase A/HEXA Monoclonal specifically detects Hexosaminidase A/HEXA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen Hexosaminidase A/HEXA
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 20F1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:3000, Flow Cytometry, ELISA
Formulation PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
Gene Accession No. NP_000511.1
Gene Alias beta-hexosaminidase subunit alpha, Beta-N-acetylhexosaminidase subunit alpha, EC 3.2.1, EC 3.2.1.52, hexosaminidase A (alpha polypeptide), Hexosaminidase subunit A, MGC99608, N-acetyl-beta-glucosaminidase subunit alpha, TSD
Gene Symbols HEXA
Host Species Mouse
Immunogen Recombinant human HEXA (89-529aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3073
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.