Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexosaminidase A/HEXA Antibody (20F1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP242629IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242629IR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242629IR Supplier Novus Biologicals Supplier No. NBP242629IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hexosaminidase A/HEXA Monoclonal specifically detects Hexosaminidase A/HEXA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen Hexosaminidase A/HEXA
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 20F1
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta-hexosaminidase subunit alpha, Beta-N-acetylhexosaminidase subunit alpha, EC 3.2.1, EC 3.2.1.52, hexosaminidase A (alpha polypeptide), Hexosaminidase subunit A, MGC99608, N-acetyl-beta-glucosaminidase subunit alpha, TSD
Gene Symbols HEXA
Host Species Mouse
Immunogen Recombinant human HEXA (89-529aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3073
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.