Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexosaminidase A/HEXA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP174127
Description
Hexosaminidase A/HEXA Polyclonal specifically detects Hexosaminidase A/HEXA in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Hexosaminidase A/HEXA | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
beta-hexosaminidase subunit alpha, Beta-N-acetylhexosaminidase subunit alpha, EC 3.2.1, EC 3.2.1.52, hexosaminidase A (alpha polypeptide), Hexosaminidase subunit A, MGC99608, N-acetyl-beta-glucosaminidase subunit alpha, TSD | |
Rabbit | |
48 kDa | |
100 μL | |
Neuroscience, Signal Transduction | |
3073 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 2-5 ug/ml | |
P06865 | |
HEXA | |
Synthetic peptides corresponding to the C terminal of HEXA. Immunizing peptide sequence: WKDFYIVEPLAFEGTPEQKALVIGGEACMWGEYVDNTNLVPRLWPRAGAV. | |
Affinity purified | |
RUO | |
Primary | |
This product is specific to Subunit or Isoform: alpha. | |
Human, Rat, Pig, Bovine, Canine, Canine, Equine, Guinea Pig, Rabbit, Sheep | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction