Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl acetate, 99%

Catalog No. p-4510080
Click to view available options
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle

Catalog No. AC148500250

Supplier: Thermo Scientific Chemicals 148500250

Only null left
Add to Cart

CAS: 142-92-7 | C8H16O2 | 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-92-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
InChI Key AOGQPLXWSUTHQB-UHFFFAOYSA-N
Synonym n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural
PubChem CID 8908
ChEBI CHEBI:87510
IUPAC Name hexyl acetate
SMILES CCCCCCOC(=O)C

Specifications

Acidity 1mg KOH/g max.
Melting Point -80.0°C
Color Colorless to Yellow
Density 0.8710g/mL
Boiling Point 168.0°C to 170.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4070 to 1.4110
Linear Formula CH3CO2C6H13
Quantity 25 mL
Beilstein 02,132
Specific Gravity 0.871
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in alcohol and ether
Formula Weight 144.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexyl acetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 205-572-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.