Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyl Methanesulfonate 96.0+%, TCI America™
SDP

Catalog No. H128225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H128225G 25 g
H12825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H128225G Supplier TCI America Supplier No. H128225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16156-50-6
Molecular Formula C7H16O3S
Molecular Weight (g/mol) 180.26
MDL Number MFCD00674707
InChI Key URIRDRHUUFRHAS-UHFFFAOYSA-N
Synonym Methanesulfonic Acid Hexyl Ester
PubChem CID 12517373
IUPAC Name hexyl methanesulfonate
SMILES CCCCCCOS(C)(=O)=O

Specifications

Color Yellow
Boiling Point 90°C
Quantity 25 g
Formula Weight 180.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyl Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.