Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hey L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | Hey L |
---|---|
Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Hey L Polyclonal antibody specifically detects Hey L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Hey L | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS (pH 7.2) and 40% Glycerol | |
26508 | |
IgG | |
Protein A purified |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
BHLHB33, bHLHb33hHeyL, Class B basic helix-loop-helix protein 33, hairy/enhancer-of-split related with YRPW motif 3, hairy/enhancer-of-split related with YRPW motif-like, hairy/enhancer-of-split related with YRPW motif-like protein, Hairy-related transcription factor 3, HEY3, HEY-like protein, hHRT3, HRT3, HRT-3, MGC12623 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MKRPKEPSGSDGESDGPIDVGQEGQLSQMARPLSTPSSSQMQARKKHRGII | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title