Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEY1 Antibody, Novus Biologicals™
SDP

Catalog No. NB396218 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396218 25 μL
NBP247436 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396218 Supplier Novus Biologicals Supplier No. NBP24743625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HEY1 Polyclonal specifically detects HEY1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen HEY1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias basic helix-loop-helix protein OAF1, BHLHb31, Cardiovascular helix-loop-helix factor 2, CHF-2, CHF2hHRT1, Class B basic helix-loop-helix protein 31, Hairy and enhancer of split-related protein 1, hairy/enhancer-of-split related with YRPW motif 1, hairy/enhancer-of-split related with YRPW motif protein 1, Hairy-related transcription factor 1, HERP2HES-related repressor protein 1, HESR-1, HESR1MGC1274, HES-related repressor protein 2, HRT1, HRT-1OAF1
Gene Symbols HEY1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: HLNNYASQREAASGAHAGLGHIPWGTVFGHHPHIAHPLLLPQNGHGNAGTTASPTEPHHQG
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 23462
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.