Learn More
HEY1 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $649.44
Specifications
| Antigen | HEY1 |
|---|---|
| Concentration | 1.06 mg/mL |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2145
![]()
|
Rockland Immunochemicals
600401HB5 |
100 μg |
Each for $649.44
|
|
|||||
50-199-2146
![]()
|
Rockland Immunochemicals
600401HB5S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human HEY1. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with the immunizing sequence to mouse and 91.7% with dog and cattle.
Hairy/enhancer-of-split related with YRPW motif protein 1 belongs to the HEY family and is a Notch Signaling Pathway Protein. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. It is a downstream effector of Notch signaling which may be required for cardiovascular development. It acts as transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. It represses transcription by the cardiac transcriptional activators GATA4 and GATA6. Hey1 is also reported to be expressed during development of the nervous system.Specifications
| HEY1 | |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q9WV93, Q9Y5J3 | |
| 15213, 23462 | |
| Primary | |
| Antibody |
| 1.06 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| AI316788; AI414254; basic helix-loop-helix protein OAF1; basic helix-loop-helix transcription factor; BC8; bHLH protein Hesr-1/Hey1; BHLHB31; BOS_13914; cardiovascular basic helix-loop-helix factor 2; Cardiovascular helix-loop-helix factor 2; CHF2; CHF-2; Class B basic helix-loop-helix protein 31; Hairy and enhancer of split-related protein 1; Hairy/E(spl)-related with YRPW motif 1; hairy/enhancer-of-split related with YRPW motif 1; Hairy/enhancer-of-split related with YRPW motif protein 1; Hairy-related transcription factor 1; Herp1; HERP2; hes related family bHLH transcription factor with YRPW motif 1; HESR1; HESR-1; hes-related family bHLH transcription factor with YRPW motif 1; HES-related repressor protein 1; HES-related repressor protein 2; hey1; hey1 protein; hHRT1; HRT1; HRT-1; id:ibd1292; LOC100344717; MGC1274; mHRT1; OAF1; zgc:110572 | |
| HEY1 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| HEY1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.