Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094155 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094155 100 μL
NB094156 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094155 Supplier Novus Biologicals Supplier No. NBP303905100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HEY2 Polyclonal specifically detects HEY2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen HEY2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias BHLHB32, bHLHb32HES-related repressor protein 2, cardiovascular basic helix-loop-helix factor 1, Cardiovascular helix-loop-helix factor 1, CHF1, Class B basic helix-loop-helix protein 32, GRLGRIDLOCK, Hairy and enhancer of split-related protein 2, hairy/enhancer-of-split related with YRPW motif 2, hairy/enhancer-of-split related with YRPW motif protein 2, Hairy-related transcription factor 2, hCHF1, HERP, HERP1hHRT2, HESR2, HESR-2, HES-related repressor protein 1, HRT2, HRT-2, MGC10720, Protein gridlock homolog
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HEY2 (NP_036391.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23493
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.