Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HGF Activator Antibody (101), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB307662 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307662 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307662 Supplier Novus Biologicals Supplier No. NBP289418APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HGF Activator Monoclonal antibody specifically detects HGF Activator in Human samples. It is validated for ELISA

Specifications

Antigen HGF Activator
Applications ELISA
Classification Monoclonal
Clone 101
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias HGF activator, HGFA, HGFAC
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human HGF Activator (Accession#: NP_001519.1; Met 1-Ser 655).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3083
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.