Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HGFR/c-MET Antibody (001), Alexa Fluor™ 700
SDP

Catalog No. NB076114 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076114 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076114 Supplier Novus Biologicals Supplier No. NBP290655AF700

Rabbit Monoclonal Antibody

HGFR/c-MET Monoclonal specifically detects HGFR/c-MET in Mouse samples. It is validated for ELISA.

Specifications

Antigen HGFR/c-MET
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias AUTS9, c-Met, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF receptor, HGF/SF receptor, HGFR, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse HGFR/c-MET (NP_032617.2; Met1-Asn929).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cancer Stem Cells, Cell Cycle and Replication, Cellular Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 4233
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.