Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HGFR/c-MET Antibody (1G7NB) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP244309 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244309 0.1 mg
NBP244309SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP244309 Supplier Novus Biologicals Supplier No. NBP244309
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Flow (Intracellular), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)

HGFR/c-MET Monoclonal specifically detects HGFR/c-MET in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen HGFR/c-MET
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone 1G7NB
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 5-10 ug/ml, Flow (Intracellular) 1:1000
Formulation PBS with No Preservative
Gene Alias AUTS9, c-Met, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF receptor, HGF/SF receptor, HGFR, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met
Gene Symbols MET
Host Species Mouse
Immunogen Partial recombinant human c-Met protein (between amino acids 170-400) [UniProt P08581].
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cancer Stem Cells, Cell Cycle and Replication, Cellular Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 4233
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.