Learn More
Description
Specifications
Specifications
| Antigen | HGFR/c-MET |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 243 |
| Conjugate | DyLight 755 |
| Dilution | Flow Cytometry, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AUTS9, c-Met, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF receptor, HGF/SF receptor, HGFR, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human HGFR/c-MET (NP_000236; Met1-Thr932). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
