Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Hh-Ag1.5

Click to view available options
Quantity:
1 mg
Description
Hh-Ag1.5 is a potent and high affinity Hedgehog (Hh) receptor agonist (EC50 = 1 nM; Ki values are 0.5 and 2.3 nM respectively, in a [3H]SAG-1.3 and [3H]Cyclopamine binding assay). It is used in protocols for the chemical reprogramming of mouse embryonic fibroblasts into neural stem cells. Also induces differentiation of hiPSCs into skin-derived precursor cells, spinal motor neurons and spinal sensory neurons.
Specifications
Specifications
Chemical Name or Material | 3-Chloro-4,7-difluoro-N-[trans-4-(methylamino)cyclohexyl]-N-[[3-(4-pyridinyl)phenyl]methyl]benzo[b]thiophene-2-carboxamide |
CAS | 612542-14-0 |
Quantity | 1 mg |
Target | Smoothened |
Molecular Formula | C28H26ClF2N3OS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction