Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HHAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094158 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094158 20 μL
NB094157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094158 Supplier Novus Biologicals Supplier No. NBP30390620UL

Rabbit Polyclonal Antibody

HHAT Polyclonal specifically detects HHAT in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen HHAT
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EC 2.3.1.-, FLJ10724, FLJ34867, GUP2, hedgehog acyltransferaserasp, MART-2, MART2sit, melanoma antigen recognized by T cells 2, Melanoma antigen recognized by T-cells 2, protein-cysteine N-palmitoyltransferase HHAT, SKI1ski, Skinny hedgehog protein 1, Skn
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HHAT (NP_060664.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55733
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.