Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HHEX Antibody (2018B), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB372969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372969 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372969 Supplier Novus Biologicals Supplier No. FAB83771L
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HHEX Monoclonal specifically detects HHEX in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.

Specifications

Antigen HHEX
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2018B
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias hematopoietically expressed homeobox, HEXHMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRHhematopoietically-expressed homeobox protein HHEX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor
Host Species Rabbit
Immunogen E. coli-derived recombinant human HHEX, Thr111-Gly270, Accession # Q03014
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 3087
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.