Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HHEX Antibody (SR1508), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322023100UL
Description
HHEX Monoclonal antibody specifically detects HHEX in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
HHEX | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
SR1508 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:40 | |
hematopoietically expressed homeobox, HEXHMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRHhematopoietically-expressed homeobox protein HHEX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor | |
A synthesized peptide derived from human HHEX (Uniprot #: Q03014) | |
100 μg | |
Developmental Biology | |
3087 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction