Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HHEX Antibody (SR1508), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | HHEX |
---|---|
Clone | SR1508 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:40 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
HHEX Monoclonal antibody specifically detects HHEX in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
HHEX | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:40 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
hematopoietically expressed homeobox, HEXHMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRHhematopoietically-expressed homeobox protein HHEX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor | |
A synthesized peptide derived from human HHEX (Uniprot #: Q03014) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1508 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Developmental Biology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3087 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title