Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hHpr1-p84-Thoc1 Antibody (5E10), Novus Biologicals™
SDP

Catalog No. NB100174 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100174 Supplier Novus Biologicals Supplier No. NB100174
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 7 publications

hHpr1-p84-Thoc1 Monoclonal specifically detects hHpr1-p84-Thoc1 in Hamster, Human, Monkey, Mouse, Rat samples. It is validated for ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen hHpr1-p84-Thoc1
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot
Classification Monoclonal
Clone 50000000000
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 2 ug/ml, Simple Western, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 0.5 - 2 ug/ml, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Gene Alias HPR1THO complex subunit 1, hTREX84, Nuclear matrix protein p84, P84, P84N5, THO complex 1, tho1
Gene Symbols THOC1
Host Species Mouse
Immunogen Amino acids 15-374 of human p84 expressed in E. coli.
Molecular Weight of Antigen 84 kDa
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9984
Test Specificity This is specific for p84.
Target Species Hamster, Human, Monkey, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.