Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hHR23b Antibody (OTI7G10), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N271269A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271269A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271269A647 Supplier Novus Biologicals Supplier No. NBP271269AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

hHR23b Monoclonal specifically detects hHR23b in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen hHR23b
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7G10
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HHR23B, HR23BUV excision repair protein RAD23 homolog B, p58, RAD23 (S. cerevisiae) homolog B, RAD23 homolog B (S. cerevisiae), RAD23, yeast homolog of, B, XP-C repair complementing complex 58 kDa, XP-C repair complementing protein, XP-C repair-complementing complex 58 kDa protein
Gene Symbols RAD23B
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-253 of human hHR23b (NP_002865) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Core ESC Like Genes, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5887.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.