Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HHV8-ORF74 Antibody (462510), PerCP, Novus Biologicals™
SDP

Catalog No. NB368245 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368245 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368245 Supplier Novus Biologicals Supplier No. FAB5264C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HHV8-ORF74 Monoclonal specifically detects HHV8-ORF74 in Virus samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen HHV8-ORF74
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 462510
Conjugate PerCP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with viral HHV8-ORF74, Met1-Thr342, Accession # AAB51506
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4961460
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.