Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ Hi Intensity LEDS for GBOX
GREENER_CHOICESDP

Catalog No. p-7220811 Shop All Synoptics, Inc. Products
Click to view available options
Color:
Green, Blue, Far Red
Green, Blue, Infa Red, Far Red
Infa Red, Far Red
Red, Green, Blue

Syngene™ Hi Intensity LEDS for GBOX includes gantry, electronics and lighting units, and works with latest GBOX ONLY (not previous modular system).

  • For use with GBOX
  • Includes gantry, electronics and lighting units

Specifications

For Use With (Application) Multiplex Western Fluorescence
Quantity 1
For Use With (Equipment) Gbox Chemi Systems
Color Green, Blue, Far Red
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.