Learn More
MilliporeSigma™ Hibar™ HPLC Columns, 125-4
Catalog No. M1501817109 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
60 SI
HPLC phase
NH2
RP18
RP8
RPSELECTB
SI3
Mobile Phase Compatibility:
NP (normal-phase)
RP (reversed-phase)
RP (when in mode) and NP
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase Mobile Phase Compatibility
M1501817109 60 SI NP (normal-phase)
M1501817141 RPSELECTB RP (reversed-phase)
M1501817076 NH2 RP (when in mode) and NP
M1501817078 HPLC phase RP (reversed-phase)
M1501817140 RP8 RP (reversed-phase)
M1501817174 SI3 NP (normal-phase)
M1501817116 RP18 RP (reversed-phase)
M1501817080 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. M1501817109 Supplier MilliporeSigma™ Supplier No. 1501817109
Add to Cart
Add to Cart
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma Hibar™ HPLC Columns, 125-4 come with customized packing (please specify packing material) and are 125mmL x 4mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 4 mm
Length (Metric) 125 mm
Mobile Phase Compatibility NP (normal-phase)
Stationary Phase 60 SI
Packing Material Customized (specify packing material)
Column Type Hibar™ HPLC Column