Learn More
MilliporeSigma™ Hibar™ HPLC Columns, 250-10
Catalog No. M1501837088 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
60 SI
DIOL5
NH2
RP18
RP8
RPSELECTB
Mobile Phase Compatibility:
NP (normal-phase)
RP (reversed-phase)
RP (when in mode) and NP
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase Mobile Phase Compatibility
M1501837088 RP8 RP (reversed-phase)
M1501837075 DIOL5 RP (when in mode) and NP
M1501837076 NH2 RP (when in mode) and NP
M1501837109 60 SI NP (normal-phase)
M1501837127 RP18 RP (reversed-phase)
M1501837094 RPSELECTB RP (reversed-phase)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. M1501837088 Supplier MilliporeSigma™ Supplier No. 1501837088
Add to Cart
Add to Cart
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma Hibar™ HPLC Columns, 250-10 come with customized packing (please specify packing material) and are 250mmL x 10mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 10 mm
Length (Metric) 250 mm
Mobile Phase Compatibility RP (reversed-phase)
Stationary Phase RP8
Packing Material Customized (specify packing material)
Column Type Hibar™ HPLC Column