Learn More
MilliporeSigma™ Hibar™ HPLC Columns, 250-4
Catalog No. M1501827101 Shop All MilliporeSigma Products
Change view
Click to view available options
Mobile Phase Compatibility:
NP (normal-phase)
RP (reversed-phase)
RP (when in mode) and NP
Stationary Phase:
100 DIOL
60 SI
NH2
RP18
RP8
RPSELECTB
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mobile Phase Compatibility Stationary Phase
M1501827101 RP (reversed-phase) -
M1501827088 RP (reversed-phase) RP8
M1501827109 NP (normal-phase) 60 SI
M1501827138 RP (reversed-phase) RP18
M1501827177 RP (when in mode) and NP NH2
M1501827075 RP (when in mode) and NP 100 DIOL
M1501827141 RP (reversed-phase) RPSELECTB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. M1501827101 Supplier MilliporeSigma™ Supplier No. 1.50182.7101
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma Hibar™ HPLC Columns, 250-4 come with customized packing (please specify packing material) and are 250mmL x 4mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 4 mm
Length (Metric) 250 mm
Mobile Phase Compatibility RP (reversed-phase)
Packing Material Customized (specify packing material)
Column Type Hibar™ HPLC Column