Learn More
MilliporeSigma™ Hibar™ HPLC Columns, 250-4.6
Catalog No. M1004247141 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
100 SI
60 SI
CN5
HPLC phase
NH2
RP18
RP8
RPSELECTB
Mobile Phase Compatibility:
NP (normal-phase)
RP (reversed-phase)
RP (when in mode) and NP
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase Mobile Phase Compatibility
M1004247141 RPSELECTB RP (reversed-phase)
M1004247071 CN5 RP (when in mode) and NP
M1004247077 NH2 RP (when in mode) and NP
M1004247078 HPLC phase RP (reversed-phase)
M1004247109 60 SI NP (normal-phase)
M1004247137 RP18 RP (reversed-phase)
M1004247220 RP8 RP (reversed-phase)
M1004247101 100 SI NP (normal-phase)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. M1004247141 Supplier MilliporeSigma™ Supplier No. 1004247141
Add to Cart
Add to Cart
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma Hibar™ HPLC Columns, 250-4.6 come with customized packing (please specify packing material) and are 250mmL x 4.6mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 4.6 mm
Length (Metric) 250 mm
Mobile Phase Compatibility RP (reversed-phase)
Stationary Phase RPSELECTB
Packing Material Customized (specify packing material)
Column Type Hibar™ HPLC Column