Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIBCH Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB352077 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB352077 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB352077 Supplier Novus Biologicals Supplier No. NBP306399APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIBCH Polyclonal antibody specifically detects HIBCH in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen HIBCH
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 3-hydroxyisobutyryl-CoA hydrolase, EC 3.1.2.4,3-hydroxyisobutyryl-coenzyme A hydrolase3-hydroxyisobutyryl-CoA hydrolase, mitochondrial, HIB-CoA hydrolase, HIBYL-CoA-H, HIBYLCOAH
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human HIBCH fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 26275
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.