Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HIC (Hydrophobic Interaction Chromatography) Media

Catalog No. p-4673268 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Capto Butyl media
Capto Phenyl (High Sub) media
Ligand Type:
Butyl
Phenyl
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Ligand Type
45-002-995 Capto Phenyl (High Sub) media Phenyl
45-002-996 Capto Butyl media Butyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-995 Supplier Cytiva Supplier No. 17545101
Only null left
Add to Cart
Add to Cart

Features high chemical stability, properties to increase productivity and epoxy coupling

  • High flow rate, high capacity and low backpressure reduce cycle times
  • Chemically stable to commonly used aqueous buffers
  • Epoxy chemical coupling

  • Capto Phenyl (high sub) has a hydrophobic ligand of phenyl and is a hydrophobic interaction chromatography medium used for protein purification
  • Capto Butyl has a hydrophobic ligand of butly and a highly cross-linked agarose matrix

Used in capture and intermediate stages of protein purification

TRUSTED_SUSTAINABILITY

Specifications

Product Type Capto Phenyl (High Sub) media
Flow Rate 600 cm/Hoμr
For Use With (Application) For Used in The Capture and Intermediate Stages of Protein PurifiCatIon
Quantity 25 mL
Ligand Type Phenyl
Media Type Highly Cross-Linked Agarose
Max. Pressure 300 kPa
Particle Size 75 μm
pH Range 3 to 13
Storage Requirements 4°C to 30°C
Storage Buffer 20% Ethanol
Binding Capacity 27 mg BSA/mL
Height (Metric) Bed 20 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.