Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIC2 Antibody (PCRP-HIC2-1B1), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB362923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362923 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362923 Supplier Novus Biologicals Supplier No. NBP314231MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIC2 Monoclonal antibody specifically detects HIC2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Protein Array

Specifications

Antigen HIC2
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunoprecipitation, Protein Array
Classification Monoclonal
Clone PCRP-HIC2-1B1
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Protein Array
Formulation 50mM Sodium Borate
Gene Alias HIC1-related gene on chromosome 22 protein, Hic-2, Hic-3, HRG22KIAA1020hic-2, hypermethylated in cancer 2, hypermethylated in cancer 2 protein, ZBTB30hic-3, Zinc finger and BTB domain-containing protein 30, ZNF907
Host Species Mouse
Immunogen Recombinant full-length human HIC2protein (Uniprot: Q96JB3)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23119
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.