Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HID1 Antibody (OTI2F4), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N272481PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272481PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272481PEC5 Supplier Novus Biologicals Supplier No. NBP272481PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HID1 Monoclonal specifically detects HID1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen HID1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2F4
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias C17orf28, chromosome 17 open reading frame 28, DMC1, DMC1FLJ43526, downregulated in multiple cancer 1, Down-regulated in multiple cancers 1, HID-1, hypothetical protein LOC283987
Gene Symbols HID1
Host Species Mouse
Immunogen Full length recombinant protein of human HID1 (NP_085133) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 283987.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.