Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Antibody (2443B) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP275977 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275977 0.1 mg
NBP275977SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP275977 Supplier Novus Biologicals Supplier No. NBP275977
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockout Validated.

Specifications

Antigen HIF-1 alpha
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay
Classification Monoclonal
Clone 2443B
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockout Validated
Formulation PBS with 0.02% Sodium Azide
Gene Alias AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, H1alpha67, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1
Gene Symbols HIF1A
Host Species Rabbit
Immunogen This recombinant HIF-1 alpha Antibody (2443B) was developed against partial recombinant human HIF-1 alpha protein (amino acids 1-826) [Uniprot# Q16665].
Purification Method Protein A or G purified
Quantity 0.1 mg
Research Discipline Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3091.0
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.