Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIF-1 alpha Antibody (2443B) - BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$243.50 - $501.50
Specifications
Antigen | HIF-1 alpha |
---|---|
Clone | 2443B |
Concentration | 1.0 mg/mL |
Dilution | Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockout Validated |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP275977SS
![]() |
Novus Biologicals
NBP275977SS |
0.025 mg |
Each for $243.50
|
|
|||||
NBP275977
![]() |
Novus Biologicals
NBP275977 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
HIF-1 alpha Monoclonal specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
HIF-1 alpha | |
1.0 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay | |
Unconjugated | |
Rabbit | |
Human | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, H1alpha67, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1 | |
HIF1A | |
IgG | |
Protein A or G purified |
2443B | |
Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
Monoclonal | |
Purified | |
Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
PBS with 0.02% Sodium Azide | |
3091.0 | |
This recombinant HIF-1 alpha Antibody (2443B) was developed against partial recombinant human HIF-1 alpha protein (amino acids 1-826) [Uniprot# Q16665]. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title